accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5445-29-4

IUPAC Name: ethyl 2-bromooctanoate
Synonyms: ethyl 2-bromooctanoate
Molecular Weight (g/mol): 251.16
Molecular Formula: C10H19BrO2
InChi Key: JIQJOKSCSVMZAN-UHFFFAOYNA-N
SMILES: CCCCCCC(Br)C(=O)OCC
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results