Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-bromooctanoate, 98+%, Thermo Scientific™

Catalog No. AAB2189614
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2189614 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2189614 Supplier Thermo Scientific Chemicals Supplier No. B2189614

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5445-29-4
Boiling Point 137°C to 140°C (25mmHg)
Flash Point 106°C (222°F)
Molecular Formula C10H19BrO2
Quantity 25g
MDL Number MFCD00042603
Beilstein 1767535
Synonym ethyl2-bromooctanoate, octanoic acid, 2-bromo-, ethyl ester, 2-bromooctanoic acid ethyl ester, ethyl-2-bromooctanoate, ethyl 2-bromocaprylate, ethyl alpha-bromocaprylate, a-bromo-octanoicacidethylester, ethyl rs-2-bromooctanoate, ksc497s8n
InChI Key JIQJOKSCSVMZAN-UHFFFAOYNA-N
SMILES CCCCCCC(Br)C(=O)OCC
IUPAC Name ethyl 2-bromooctanoate
Molecular Weight (g/mol) 251.16
PubChem CID 95580
Formula Weight 251.17
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-bromooctanoate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.