accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54458-61-6

IUPAC Name: 2,3,4,5-tetramethylcyclopent-2-en-1-one
Synonyms: 2,3,4,5-tetramethylcyclopent-2-en-1-one
Molecular Weight (g/mol): 138.21
Molecular Formula: C9H14O
InChi Key: ARUAYSANQMCCEN-UHFFFAOYNA-N
SMILES: CC1C(C)C(=O)C(C)=C1C
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)