Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetramethyl-2-cyclopentenone, cis + trans, 95%

Catalog No. AAL0961714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0961714 25 g
AAL0961706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0961714 Supplier Thermo Scientific Chemicals Supplier No. L0961714
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 54458-61-6 | C9H14O | 138.21 g/mol

2,3,4,5-Tetramethyl-2-cyclopentenone was used in the synthesis of chiral pre-ligands, (R)-3,3?-bis(tetramethylcyclopentadienyl)-2,2?-bismethoxy-1,1?-bisnaphthalene and (R)-3-tetramethylcyclopentadienyl-2,2?-bismethoxy-1,1?-bisnaphthalene by reacting with (R)-3,3?-dilithium-2,2?-bismethoxy-1,1?-bisnaphthalene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5-Tetramethyl-2-cyclopentenone was used in the synthesis of chiral pre-ligands, (R)-3,3′-bis(tetramethylcyclopentadienyl)-2,2′-bismethoxy-1,1′-bisnaphthalene and (R)-3-tetramethylcyclopentadienyl-2,2′-bismethoxy-1,1′-bisnaphthalene by reacting with (R)-3,3′-dilithium-2,2′-bismethoxy-1,1′-bisnaphthalene.

Solubility
Not miscible with water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54458-61-6
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00010248
InChI Key ARUAYSANQMCCEN-UHFFFAOYSA-N
Synonym 2,3,4,5-tetramethylcyclopent-2-enone, 2,3,4,5-tetramethyl-2-cyclopentenone, 2,3,4,5-tetramethyl-2-cyclopenten-1-one, 2-cyclopenten-1-one, 2,3,4,5-tetramethyl, 2,3,4,5-tetramethyl-2-cyclopentenone, cis + trans, tetramethylcyclopent-2-enone, pubchem15060, acmc-1asp4, 2,3,4,5-tetramethylcyclopentenone, 2,3,4,5-tetramethyl-cyclopent-2-enone
PubChem CID 41092
IUPAC Name 2,3,4,5-tetramethylcyclopent-2-en-1-one
SMILES CC1C(C(=O)C(=C1C)C)C

Specifications

Density 0.917
Boiling Point 99°C to 100°C (30 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.476
Quantity 25 g
Beilstein 2324088
Solubility Information Not miscible with water.
Formula Weight 138.21
Percent Purity 95%
Chemical Name or Material 2,3,4,5-Tetramethyl-2-cyclopentenone, Cis + Trans
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.