accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5448-47-5

IUPAC Name: 2-(1H-indol-3-yl)acetohydrazide
Synonyms: 3-indoleacetic acid hydrazide
Molecular Weight (g/mol): 189.22
Molecular Formula: C10H11N3O
InChi Key: GYHLCXMCGCVVCG-UHFFFAOYSA-N
SMILES: NNC(=O)CC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)