Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-3-acetic acid hydrazide, 98%, Thermo Scientific™

Catalog No. AAA1656206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1656206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1656206 Supplier Thermo Scientific Chemicals Supplier No. A1656206

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5448-47-5
Molecular Formula C10H11N3O
Molecular Weight (g/mol) 189.218
MDL Number MFCD00005634
InChI Key GYHLCXMCGCVVCG-UHFFFAOYSA-N
Synonym 2-1h-indol-3-yl acetohydrazide, indole-3-acetic acid hydrazide, 3-indoleacetic acid hydrazide, 1-3-indolylacetyl hydrazine, 3-indolylacetic acid hydrazide, 1h-indole-3-acetic acid, hydrazide, hydrazine, 1-3-indolylacetyl, indole 3-acethydrazide, indole-3-acetic hydrazide, indole-3-acetic acid, hydrazide
PubChem CID 21589
IUPAC Name 2-(1H-indol-3-yl)acetohydrazide
SMILES C1=CC=C2C(=C1)C(=CN2)CC(=O)NN

Specifications

Melting Point 142°C to 145°C
Quantity 5 g
Formula Weight 189.22
Percent Purity 98%
Chemical Name or Material Indole-3-acetic acid hydrazide
EINECSNumber 226-672-7
RTECSNumber NL3600000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.