accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHNNNH2

CAS RN 545380-34-5

IUPAC Name: N4-[2-(4-phenoxyphenyl)ethyl]quinazoline-4,6-diamine
Synonyms: N4-[2-(4-phenoxyphenyl)ethyl]quinazoline-4,6-diamine
Molecular Weight (g/mol): 356.43
Molecular Formula: C22H20N4O
InChi Key: IBAKVEUZKHOWNG-UHFFFAOYSA-N
SMILES: NC1=CC2=C(NCCC3=CC=C(OC4=CC=CC=C4)C=C3)N=CN=C2C=C1
Product Type 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results