accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBr

CAS RN 54550-36-6

IUPAC Name: 1-bromo-2-(2-ethoxyethoxy)ethane
Synonyms: 1-bromo-2-(2-ethoxyethoxy)ethane
Molecular Weight (g/mol): 197.07
Molecular Formula: C6H13BrO2
InChi Key: UEDIWIFQWRXXJG-UHFFFAOYSA-N
SMILES: CCOCCOCCBr
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)