CAS RN 546-79-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sabinene hydrate, ≥97% (GC), MilliporeSigma™ Supelco™
Sabinene hydrate is a monoterpene present in Origanum majorana, which is synthesized by the enzyme sabinene hydrate synthase.