accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5460-09-3

IUPAC Name: sodium 5-amino-4-hydroxy-7-sulfonaphthalene-2-sulfonate
Synonyms: sodium 5-amino-4-hydroxy-7-sulfonaphthalene-2-sulfonate
Molecular Weight (g/mol): 341.28
Molecular Formula: C10H8NNaO7S2
InChi Key: QPILZZVXGUNELN-UHFFFAOYSA-M
SMILES: [Na+].NC1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)