Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt monohydrate, 80%, Thermo Scientific™

Catalog No. AC400842500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC400842500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC400842500 Supplier Thermo Scientific Chemicals Supplier No. 400842500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5460-09-3
Molecular Formula C10H8NNaO7S2
Molecular Weight (g/mol) 341.28
InChI Key QPILZZVXGUNELN-UHFFFAOYSA-M
Synonym h acid sodium, 1-amino-8-naphthol-3,6-disulfonic acid monosodiumsalt, 4-amino-5-hydroxynaphthalene-2,7-disulfonic acid 2-sodium salt
PubChem CID 102116466
IUPAC Name sodium;8-amino-3,6-disulfonaphthalen-1-olate
SMILES C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)N)[O-])S(=O)(=O)O.[Na+]

Specifications

Quantity 250 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 359.32
Chemical Name or Material 8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.