accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3OH

CAS RN 54630-50-1

IUPAC Name: 5-methylheptan-2-ol
Synonyms: 2-heptanol, 5-methyl-
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: FYMBAYNKBWGEIK-UHFFFAOYNA-N
SMILES: CCC(C)CCC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results