Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-heptanol, TCI America™
SDP

Catalog No. M12225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12225ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12225ML Supplier TCI America Supplier No. M12225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54630-50-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00060896
InChI Key FYMBAYNKBWGEIK-UHFFFAOYSA-N
PubChem CID 41143
IUPAC Name 5-methylheptan-2-ol
SMILES CCC(C)CCC(C)O

Specifications

Color Colorless
Boiling Point 172°C
Quantity 5 mL
UN Number 1987
Formula Weight 130.23
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-heptanol
EINECSNumber (2)-0217
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.