accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54649-21-7

IUPAC Name: furan-3-carboximidamide hydrochloride
Synonyms: furan-3-carboximidamide hydrochloride
Molecular Weight (g/mol): 146.57
Molecular Formula: C5H7ClN2O
InChi Key: RJLCJJIXGGSRPZ-UHFFFAOYSA-N
SMILES: Cl.NC(=N)C1=COC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results