accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5465-09-8

IUPAC Name: 3,4-dimethylcyclohexan-1-one
Synonyms: 3,4-dimethylcyclohexan-1-one
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: ZDCYWXYPRPCJOY-UHFFFAOYNA-N
SMILES: CC1CCC(=O)CC1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results