Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylcyclohexanone (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. D128110ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D128110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D128110ML Supplier TCI America Supplier No. D128110ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5465-09-8
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00045552
InChI Key ZDCYWXYPRPCJOY-UHFFFAOYNA-N
PubChem CID 138519
IUPAC Name 3,4-dimethylcyclohexan-1-one
SMILES CC1CCC(=O)CC1C

Specifications

Color Yellow
Quantity 10 mL
UN Number 1224
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethylcyclohexanone (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.