accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOCH3(E/Z)

CAS RN 5466-77-3

IUPAC Name: 2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
Synonyms: 2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 290.4
Molecular Formula: C18H26O3
InChi Key: YBGZDTIWKVFICR-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)