Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octinoxate, USP, 95-105%, Spectrum™ Chemical
SDP

Catalog No. 18604866 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-866 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-866 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. O11232.5KG

missing translation for 'validMainframeMsg'

O1123, 5466-77-3, C18H26O3

Spectrum™ Chemical Octinoxate, USP absorbs ultraviolet radiation in the UVB range and is used in sunscreens and other cosmetics to protect from skin damage. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5466-77-3
Molecular Formula C18H26O3
Molecular Weight (g/mol) 290.40
MDL Number MFCD00072582
InChI Key YBGZDTIWKVFICR-UHFFFAOYNA-N
IUPAC Name 2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
SMILES CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1

Specifications

Acidity To pass test
CAS Max % 1
Packaging Amber Glass Bottle
Refractive Index 1.542-1.548
Quantity 2.5 kg
Specific Gravity 1.005-1.013
Percent Purity 95 to 105%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.