CAS RN 5466-89-7
IUPAC Name:
1-(3,4-dihydroxyphenyl)-2-methylpropan-1-one
Synonyms:
1-(3,4-dihydroxyphenyl)-2-methylpropan-1-one
Molecular Weight (g/mol):
180.2
Molecular Formula:
C10H12O3
InChi Key:
VDQLKIBLTMPAHI-UHFFFAOYSA-N
SMILES:
CC(C)C(=O)C1=CC=C(O)C(O)=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals U-0521, 98%
CAS: 5466-89-7 Molecular Formula: C10H12O3 Synonym: 3',4'-Dihydroxy-2-methylpropiophenone;