accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54660-78-5

IUPAC Name: 4-chloropyrimidin-5-amine
Synonyms: 4-chloropyrimidin-5-amine
Molecular Weight (g/mol): 129.55
Molecular Formula: C4H4ClN3
InChi Key: LHGMCUVJFRBVBH-UHFFFAOYSA-N
SMILES: NC1=CN=CN=C1Cl
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)