CAS RN 5467-57-2
IUPAC Name:
2-chloroquinoline-4-carboxylate
Synonyms:
2-chloroquinoline-4-carboxylate
Molecular Weight (g/mol):
206.61
Molecular Formula:
C10H5ClNO2
InChi Key:
ICNCOMYUODLTAI-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=C2C=CC=CC2=NC(Cl)=C1
1
–
1
of
1
results
1
2-Chloroquinoline-4-carboxylic Acid 97.0+%, TCI America™
CAS: 5467-57-2 Molecular Formula: C10H5ClNO2 Molecular Weight (g/mol): 206.61 MDL Number: MFCD00023940 InChI Key: ICNCOMYUODLTAI-UHFFFAOYSA-M PubChem CID: 230582 IUPAC Name: 2-chloroquinoline-4-carboxylate SMILES: [O-]C(=O)C1=C2C=CC=CC2=NC(Cl)=C1