Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroquinoline-4-carboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. C17035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17035G 5 g
C170325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17035G Supplier TCI America Supplier No. C17035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5467-57-2
Molecular Formula C10H5ClNO2
Molecular Weight (g/mol) 206.61
MDL Number MFCD00023940
InChI Key ICNCOMYUODLTAI-UHFFFAOYSA-M
PubChem CID 230582
IUPAC Name 2-chloroquinoline-4-carboxylate
SMILES [O-]C(=O)C1=C2C=CC=CC2=NC(Cl)=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 207.61
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroquinoline-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.