CAS RN 54673-07-3
CAS RN 54673-07-3
IUPAC Name:
(2E)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoate
Synonyms:
(2E)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoate
Molecular Weight (g/mol):
188.16
Molecular Formula:
C10H6NO3
InChi Key:
HPLNTJVXWMJLNJ-XBXARRHUSA-M
SMILES:
OC1=CC=CC(\C=C(/C#N)C([O-])=O)=C1
1
–
1
of
1
results
1
alpha-Cyano-3-hydroxycinnamic Acid 98.0+%, TCI America™
CAS: 54673-07-3 Molecular Formula: C10H7NO3 Molecular Weight (g/mol): 189.17 MDL Number: MFCD00004203 InChI Key: HPLNTJVXWMJLNJ-YWEYNIOJSA-N Synonym: 2z-2-cyano-3-3-hydroxyphenyl prop-2-enoic acid PubChem CID: 21338270 IUPAC Name: (Z)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoic acid SMILES: C1=CC(=CC(=C1)O)C=C(C#N)C(=O)O