accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5468-71-3

IUPAC Name: 3,5-dichloropyridine-2-carboxamide
Synonyms: 3,5-dichloropyridine-2-carboxamide
Molecular Weight (g/mol): 191.01
Molecular Formula: C6H4Cl2N2O
InChi Key: QIRSAVRBVNYEMF-UHFFFAOYSA-N
SMILES: NC(=O)C1=NC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results