accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3BrH3C

CAS RN 5469-19-2

IUPAC Name: 1-bromo-2,4,5-trimethylbenzene
Synonyms: 1-bromo-2,4,5-trimethylbenzene
Molecular Weight (g/mol): 199.09
Molecular Formula: C9H11Br
InChi Key: SCZXFZRJDVZMJI-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(Br)C=C1C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)