Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1,2,4-trimethylbenzene, 99%, Thermo Scientific™

Catalog No. AAB2327122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2327122 100 g
AAB2327114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2327122 Supplier Thermo Scientific Chemicals Supplier No. B2327122

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5469-19-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00000072
InChI Key SCZXFZRJDVZMJI-UHFFFAOYSA-N
PubChem CID 79610
IUPAC Name 1-bromo-2,4,5-trimethylbenzene
SMILES CC1=CC(=C(C=C1C)Br)C

Specifications

Melting Point 71°C to 73°C
Boiling Point 233°C to 235°C
Quantity 100 g
Synonym 5-bromo-1,2,4-trimethylbenzene, 5-bromopseudocumene, benzene, 1-bromo-2,4,5-trimethyl, acmc-1art4, 2,4,5-trimethylbromobenzene, 5-bromo-1,2,4-trimethyl-benzene, #, benzene,1-bromo-2,4,5-trimethyl
Formula Weight 199.09
Percent Purity 99%
Chemical Name or Material 5-Bromo-1,2,4-trimethylbenzene
EINECSNumber 226-793-5
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.