CAS RN 5469-26-1
1-Bromopinacolone, 97+%
CAS: 5469-26-1 Molecular Formula: C6H11BrO Molecular Weight (g/mol): 179.057 MDL Number: MFCD00000206 InChI Key: SAIRZMWXVJEBMO-UHFFFAOYSA-N Synonym: 1-bromopinacolone,bromopinacolone,1-bromo-3,3-dimethyl-2-butanone,1-bromopinacolin,bromopinacolin,bromomethyl tert-butyl ketone,pivaloylmethyl bromide,tert-butyl bromomethyl ketone,2-butanone, 1-bromo-3,3-dimethyl,omega-brompinakolin german PubChem CID: 21642 IUPAC Name: 1-bromo-3,3-dimethylbutan-2-one SMILES: CC(C)(C)C(=O)CBr
1-Bromopinacolone, 93%
CAS: 5469-26-1 Molecular Formula: C6H11BrO Molecular Weight (g/mol): 179.06 MDL Number: MFCD00000206 InChI Key: SAIRZMWXVJEBMO-UHFFFAOYSA-N Synonym: 1-bromopinacolone,bromopinacolone,1-bromo-3,3-dimethyl-2-butanone,1-bromopinacolin,bromopinacolin,bromomethyl tert-butyl ketone,pivaloylmethyl bromide,tert-butyl bromomethyl ketone,2-butanone, 1-bromo-3,3-dimethyl,omega-brompinakolin german PubChem CID: 21642 IUPAC Name: 1-bromo-3,3-dimethylbutan-2-one SMILES: CC(C)(C)C(=O)CBr
1-Bromo-3,3-dimethyl-2-butanone 95.0+%, TCI America™
CAS: 5469-26-1 Molecular Formula: C6H11BrO Molecular Weight (g/mol): 179.057 MDL Number: MFCD00000206 InChI Key: SAIRZMWXVJEBMO-UHFFFAOYSA-N Synonym: 1-bromopinacolone,bromopinacolone,1-bromo-3,3-dimethyl-2-butanone,1-bromopinacolin,bromopinacolin,bromomethyl tert-butyl ketone,pivaloylmethyl bromide,tert-butyl bromomethyl ketone,2-butanone, 1-bromo-3,3-dimethyl,omega-brompinakolin german PubChem CID: 21642 IUPAC Name: 1-bromo-3,3-dimethylbutan-2-one SMILES: CC(C)(C)C(=O)CBr
Sigma Aldrich 1-Bromopinacolone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 1-Bromopinacolone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
1-Bromo-3,3-dimethylbutan-2-one, Tech., Thermo Scientific™
CAS: 5469-26-1 Molecular Formula: C6H11BrO Molecular Weight (g/mol): 179.057 MDL Number: MFCD00000206 InChI Key: SAIRZMWXVJEBMO-UHFFFAOYSA-N Synonym: 1-bromopinacolone,bromopinacolone,1-bromo-3,3-dimethyl-2-butanone,1-bromopinacolin,bromopinacolin,bromomethyl tert-butyl ketone,pivaloylmethyl bromide,tert-butyl bromomethyl ketone,2-butanone, 1-bromo-3,3-dimethyl,omega-brompinakolin german PubChem CID: 21642 IUPAC Name: 1-bromo-3,3-dimethylbutan-2-one SMILES: CC(C)(C)C(=O)CBr