accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOClN

CAS RN 5470-22-4

IUPAC Name: 4-chloropyridine-2-carboxylate
Synonyms: 4-chloropyridine-2-carboxylate
Molecular Weight (g/mol): 156.55
Molecular Formula: C6H3ClNO2
InChi Key: NNMYRMGMVLMQAY-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(Cl)=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)