Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloropyridine-2-carboxylic acid, 95%, Thermo Scientific™

Catalog No. AC454430010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454430010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454430010 Supplier Thermo Scientific Chemicals Supplier No. 454430010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5470-22-4
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
InChI Key NNMYRMGMVLMQAY-UHFFFAOYSA-N
Synonym 4-chloropicolinic acid, 4-chloro-pyridine-2-carboxylic acid, 4-chloro-2-pyridinecarboxylic acid, 4-chloro-2-picolinic acid, 2-pyridinecarboxylic acid, 4-chloro, 4-chloro picolinic acid, 2-carboxy-4-chloropyridine, chloropicolinicacid, pubchem12731, acmc-1bn7z
PubChem CID 230890
IUPAC Name 4-chloropyridine-2-carboxylic acid
SMILES C1=CN=C(C=C1Cl)C(=O)O

Specifications

Melting Point 180.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 157.56
Percent Purity 95%
Chemical Name or Material 4-Chloropyridine-2-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.