accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3ONCl

CAS RN 547740-43-2

IUPAC Name: 2-(tert-butoxy)-6-chloropyridine
Synonyms: 2-(tert-butoxy)-6-chloropyridine
Molecular Weight (g/mol): 185.65
Molecular Formula: C9H12ClNO
InChi Key: PLDRTFJRLUKTTA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC1=NC(Cl)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)