accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHO

CAS RN 54857-86-2

IUPAC Name: 5-(tetradecyloxy)furan-2-carboxylic acid
Synonyms: 5-(tetradecyloxy)furan-2-carboxylic acid
Molecular Weight (g/mol): 324.46
Molecular Formula: C19H32O4
InChi Key: CZRCFAOMWRAFIC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCOC1=CC=C(O1)C(O)=O
Product Type 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results