CAS RN 54881-49-1
IUPAC Name:
2-chloro-3-methoxybenzaldehyde
Synonyms:
2-chloro-3-methoxybenzaldehyde
Molecular Weight (g/mol):
170.59
Molecular Formula:
C8H7ClO2
InChi Key:
XKLMLKSXPIITAL-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(C=O)=C1Cl
1
–
1
of
1
results
1
2-Chloro-3-methoxybenzaldehyde 98.0+%, TCI America™
CAS: 54881-49-1 Molecular Formula: C8H7ClO2 Molecular Weight (g/mol): 170.592 MDL Number: MFCD07369787 InChI Key: XKLMLKSXPIITAL-UHFFFAOYSA-N PubChem CID: 12575695 IUPAC Name: 2-chloro-3-methoxybenzaldehyde SMILES: COC1=CC=CC(=C1Cl)C=O