Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-methoxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. C21855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21855G 5 g
C218525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21855G Supplier TCI America Supplier No. C21855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54881-49-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD07369787
InChI Key XKLMLKSXPIITAL-UHFFFAOYSA-N
PubChem CID 12575695
IUPAC Name 2-chloro-3-methoxybenzaldehyde
SMILES COC1=CC=CC(=C1Cl)C=O

Specifications

Melting Point 53°C
Quantity 5 g
UN Number 3077
Formula Weight 170.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-methoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.