accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54882-04-1

IUPAC Name: ethyl 2-(benzenesulfinyl)acetate
Synonyms: ethyl 2-(benzenesulfinyl)acetate
Molecular Weight (g/mol): 212.26
Molecular Formula: C10H12O3S
InChi Key: HVVJMTVECSPOGD-UHFFFAOYNA-N
SMILES: CCOC(=O)CS(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results