Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Phenylsulfinylacetate 85.0+%, TCI America™
SDP

Catalog No. P11355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11355G Supplier TCI America Supplier No. P11355G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54882-04-1
Molecular Formula C10H12O3S
Molecular Weight (g/mol) 212.263
MDL Number MFCD00191677
InChI Key HVVJMTVECSPOGD-UHFFFAOYSA-N
Synonym Phenylsulfinylacetic Acid Ethyl Ester
PubChem CID 10932761
IUPAC Name ethyl 2-(benzenesulfinyl)acetate
SMILES CCOC(=O)CS(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 154°C
Quantity 5 g
Formula Weight 212.26
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Phenylsulfinylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.