accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 549-18-8

IUPAC Name: hydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
Synonyms: amitriptyline hydrochloride
Molecular Weight (g/mol): 313.87
Molecular Formula: C20H24ClN
InChi Key: KFYRPLNVJVHZGT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2CCC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (5)
  • (5)
Percent Purity 
  • (1)
  • (1)
  • (5)
  • (5)
  • (2)
Grade 
  • (2)
  • (10)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (10)
  • (5)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (5)
  • (1)
  • (4)
  • (5)
  • (5)
  • (1)
  • (1)
  • (5)
  • (5)
  • (2)
  • (2)
  • (10)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)