Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Amitriptyline Hydrochloride, USP
SDP

Catalog No. 14186123 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-123 5 g
14-185-612 1 kg
14-185-446 100 g
14-185-162 25 g
14-185-156 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-186-123 Supplier Medisca Supplier No. 018903
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amitriptyline Hydrochloride USP ranges from white to practically white crystalline powder or small crystals. It is odorless to practically odorless.

Amitriptyline Hydrochloride USP ranges from white to practically white crystalline powder or small crystals. It is odorless to practically odorless. Protect from direct sunlight and moisture. Freely soluble in water and in alcohol.

Specifications

Chemical Name or Material Amitriptyline Hydrochloride, USP
Packaging Jar 30ml, White, HDPE, Round, 28-400, TRIVENI-DB, CT
CAS 549-18-8
Type Drug Standard
Molecular Weight (g/mol) 313.9
Quantity 5 g
Percent Purity 98 to 102 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.