accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54947-34-1

IUPAC Name: (6E)-5-amino-6-(N-hydroxyimino)hexane-1,2,3,4-tetrol hydrochloridyl
Synonyms: (6E)-5-amino-6-(N-hydroxyimino)hexane-1,2,3,4-tetrol hydrochloridyl
Molecular Weight (g/mol): 229.64
Molecular Formula: C6H14ClN2O5
InChi Key: CWOXXORKGVTLAA-FOBNBMCVNA-N
SMILES: [Cl].NC(\C=N\O)C(O)C(O)C(O)CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results