Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucosamine Oxime Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. G00451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G00451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G00451G Supplier TCI America Supplier No. G00451G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54947-34-1
Molecular Formula C6H14ClN2O5
Molecular Weight (g/mol) 229.64
MDL Number MFCD00060165
InChI Key CWOXXORKGVTLAA-FOBNBMCVNA-N
PubChem CID 77519621
IUPAC Name (6E)-5-amino-6-(N-hydroxyimino)hexane-1,2,3,4-tetrol hydrochloridyl
SMILES [Cl].NC(\C=N\O)C(O)C(O)C(O)CO

Specifications

Color White
Quantity 1 g
Formula Weight 230.65
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material D-Glucosamine Oxime Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.