accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CNH3CCH3OOCH3

CAS RN 549495-32-1

IUPAC Name: 4-methoxy-N,N-bis(2-methylpropyl)benzamide
Synonyms: 4-methoxy-N,N-bis(2-methylpropyl)benzamide
Molecular Weight (g/mol): 263.38
Molecular Formula: C16H25NO2
InChi Key: NGYLTVYQZBPXBN-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)N(CC(C)C)CC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results