Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diisobutyl-4-methoxybenzamide, 97%, Thermo Scientific™

Catalog No. AAH59472MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59472MD 250 mg
AAH5947203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59472MD Supplier Thermo Scientific Chemicals Supplier No. H59472MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 549495-32-1
Molecular Formula C16H25NO2
Molecular Weight (g/mol) 263.381
MDL Number MFCD01011473
InChI Key NGYLTVYQZBPXBN-UHFFFAOYSA-N
Synonym 4-methoxy-n,n-bis 2-methylpropyl benzamide, n,n-diisobutyl-4-methoxybenzamide
PubChem CID 4056518
IUPAC Name 4-methoxy-N,N-bis(2-methylpropyl)benzamide
SMILES CC(C)CN(CC(C)C)C(=O)C1=CC=C(C=C1)OC

Specifications

Quantity 250 mg
Formula Weight 263.38
Percent Purity 97%
Chemical Name or Material N,N-Diisobutyl-4-methoxybenzamide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.