accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHCl

CAS RN 54954-73-3

IUPAC Name: methyl 1-azabicyclo[2.2.2]octane-3-carboxylate hydrochloride
Synonyms: methyl 1-azabicyclo[2.2.2]octane-3-carboxylate hydrochloride
Molecular Weight (g/mol): 205.68
Molecular Formula: C9H16ClNO2
InChi Key: NDTQASSGEAZYOT-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C1CN2CCC1CC2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)