Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-quinuclidinecarboxylate hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. AAL19132MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19132MD 250 mg
AAL1913203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19132MD Supplier Thermo Scientific Chemicals Supplier No. L19132MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 54954-73-3 | C9H16ClNO2 | 205.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54954-73-3
Molecular Formula C9H16ClNO2
Molecular Weight (g/mol) 205.68
MDL Number MFCD01073582
InChI Key NDTQASSGEAZYOT-UHFFFAOYNA-N
Synonym methyl 3-quinuclidinecarboxylate hydrochloride, methyl quinuclidine-3-carboxylate hydrochloride, quinuclidine-3-carboxylic acid methyl ester hydrochloride, methyl 1-azabicyclo 2.2.2 octane-3-carboxylate hydrochloride, acmc-1ao36, methyl 1-azabicyclo 2.2.2 octane-3-carboxylate-hydrogen chloride 1/1
PubChem CID 24721307
IUPAC Name methyl 1-azabicyclo[2.2.2]octane-3-carboxylate;hydrochloride
SMILES Cl.COC(=O)C1CN2CCC1CC2

Specifications

Melting Point ∽160°C (decomposition)
Beilstein 4272456
Quantity 250 mg
Formula Weight 205.69
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material Methyl 3-quinuclidinecarboxylate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.