CAS RN 54960-65-5
IUPAC Name:
4-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}butane-1-sulfonic acid
Synonyms:
4-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}butane-1-sulfonic acid
Molecular Weight (g/mol):
257.3
Molecular Formula:
C8H19NO6S
InChi Key:
VFTZCDVTMZWNBF-UHFFFAOYSA-N
SMILES:
OCC(CO)(CO)NCCCCS(O)(=O)=O
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals TABS, 99%
Thermo Scientific Chemicals TABS, 99%
CAS: 54960-65-5 Molecular Weight (g/mol): 257.301 Synonym: N-Tris(hydroxymethyl)methyl-4-aminobutanesulfonic acid