accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55-03-8

NaNH2(S)IOIOHIIOO

CAS RN 55-03-8

IUPAC Name: sodium (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoate
Synonyms: levoxyl eltroxin thyrax euthyrox L-thyroxine sodium levotiroxina de sodio novothyrox letter sodium L-thyroxine levothyroxine sodium anhydrous
Molecular Weight (g/mol): 798.86
Molecular Formula: C15H10I4NNaO4
InChi Key: YDTFRJLNMPSCFM-YDALLXLXSA-M
SMILES: [Na+].N[C@@H](CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)