Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Thyroxine Sodium Salt Pentahydrate 98.0+%, TCI America™
SDP

Catalog No. T02451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T02451G 1 g
T0245100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T02451G Supplier TCI America Supplier No. T02451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55-03-8
Molecular Formula C15H10I4NNaO4
Molecular Weight (g/mol) 798.856
MDL Number MFCD00002596
InChI Key YDTFRJLNMPSCFM-YDALLXLXSA-M
Synonym (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionic Acid Sodium Salt, Sodium (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionate
PubChem CID 23666112
ChEBI CHEBI:6446
IUPAC Name sodium;(2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoate
SMILES C1=C(C=C(C(=C1I)OC2=CC(=C(C(=C2)I)O)I)I)CC(C(=O)[O-])N.[Na+]

Specifications

Quantity 1 g
UN Number 2811
Formula Weight 798.86
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Thyroxine Sodium Salt Pentahydrate
RTECSNumber XP3583000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.