accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH2(S)(R)OH(S)OH(R)OHHOOHOH

CAS RN 5505-63-5

IUPAC Name: hydrogen (2S,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanoic acid chloride
Synonyms: hydrogen D-mannosaminic acid chloride
Molecular Weight (g/mol): 231.63
Molecular Formula: C6H14ClNO6
InChi Key: JMVMHPCPPSZGNY-BZWNWTOPSA-N
SMILES: [H+].[Cl-].N[C@@H]([C@@H](O)[C@H](O)[C@H](O)CO)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results