accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3CH3CH3OHOH3CH3C(E)(E)(E)(E)

CAS RN 55079-83-9

IUPAC Name: (2E,4E,6E,8E)-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
Synonyms: acitretin acitretin - chemical acitretinum andreu brand of acitretin 13-cis-acitretin soriatane acitretin, (z,e,e,e)-isomer all-trans-acitretin (z,e,e,e)-acitretin 13-cis-etretin
Molecular Weight (g/mol): 326.44
Molecular Formula: C21H26O3
InChi Key: IHUNBGSDBOWDMA-AQFIFDHZSA-N
SMILES: COC1=CC(C)=C(\C=C\C(\C)=C\C=C\C(\C)=C\C(O)=O)C(C)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results