accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 5518-09-2

IUPAC Name: 2-(naphthalen-1-yl)propanedinitrile
Synonyms: 2-(naphthalen-1-yl)propanedinitrile
Molecular Weight (g/mol): 192.22
Molecular Formula: C13H8N2
InChi Key: JSYNLGSYUCZAGV-UHFFFAOYSA-N
SMILES: N#CC(C#N)C1=CC=CC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results