Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthylmalononitrile 99.0+%, TCI America™
SDP

Catalog No. N05501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N05501G Supplier TCI America Supplier No. N05501G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5518-09-2
Molecular Formula C13H8N2
Molecular Weight (g/mol) 192.22
MDL Number MFCD00142672
InChI Key JSYNLGSYUCZAGV-UHFFFAOYSA-N
Synonym 1-(Dicyanomethyl)naphthalene
PubChem CID 10910363
IUPAC Name 2-naphthalen-1-ylpropanedinitrile
SMILES C1=CC=C2C(=C1)C=CC=C2C(C#N)C#N

Specifications

Melting Point 166°C
Quantity 1 g
Formula Weight 192.22
Percent Purity ≥99.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthylmalononitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.